2-(4-Methoxyphenyl)-5,5-dimethyl-1,3,2-dioxaborinane
Supplier: Apollo Scientific
OR310982-5G
APOSOR310982-5GEA 72.5 EUR
APOSOR310982-5G
2-(4-Methoxyphenyl)-5,5-dimethyl-1,3,2-dioxaborinane
2-(4-Methoxyphenyl)-5,5-dimethyl-1,3,2-dioxaborinane
Formula: C₁₂H₁₇BO₃
MW: 220.073 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09909078
CAS Number: 213596-33-9