2-[(4-Methoxyphenyl)methyl]-3,5-dioxo-2,9-diazaspiro[5.5]undecan-9-yl 2,2-dimethylpropanoate
Supplier: Apollo Scientific
OR311070-1G OR311070-500MG
APOSOR311070-1GEA 1230 EUR
APOSOR311070-1G APOSOR311070-500MG
2-[(4-Methoxyphenyl)methyl]-3,5-dioxo-2,9-diazaspiro[5.5]undecan-9-yl 2,2-dimethylpropanoate
2-[(4-Methoxyphenyl)methyl]-3,5-dioxo-2,9-diazaspiro[5.5]undecan-9-yl 2,2-dimethylpropanoate
Formula: C₂₂H₃₀N₂O₅
Storage Temperature: Ambient
MDL Number: MFCD28122607
CAS Number: 1312814-90-6