Cyclobutane-1,1-diylbis(methylene) bis(4-methylbenzenesulphonate)
Supplier: Apollo Scientific
OR312193-1G OR312193-5G
APOSOR312193-1GEA 480 EUR
APOSOR312193-1G APOSOR312193-5G
Cyclobutane-1,1-diylbis(methylene) bis(4-methylbenzenesulphonate)
Cyclobutane-1,1-diylbis(methylene) bis(4-methylbenzenesulphonate)
Formula: C₂₀H₂₄O₆S₂
MW: 424.531 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22418996
CAS Number: 22308-09-4
UN: 2585
ADR: 8 III