2,5-Diazaspiro[3.5]nonane, N2-BOC protected
Supplier: Apollo Scientific
Warning
Synonyms: 2-(tert-Butoxycarbonyl)-2,5-diazaspiro[3.5]nonane, tert-Butyl 2,5-diazaspiro[3.5]nonane-2-carboxylate
OR312203-1G OR312203-250MG OR312203-500MG
APOSOR312203-1GEA 665 EUR
APOSOR312203-1G APOSOR312203-250MG APOSOR312203-500MG
2,5-Diazaspiro[3.5]nonane, N2-BOC protected
2,5-Diazaspiro[3.5]nonane, N2-BOC protected
Formula: C₁₂H₂₂N₂O₂
MW: 226.315 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14581200
CAS Number: 1246034-93-4