tert-Butyl (3-aminobicyclo[1.1.1]pent-1-yl)carbamate
Supplier: Apollo Scientific
Warning
Synonyms: tert-Butyl (3-aminobicyclo[1.1.1]pentan-1-yl)carbamate
OR312435-100MG OR312435-1G OR312435-250MG
APOSOR312435-100MGEA 95.7 EUR
APOSOR312435-100MG APOSOR312435-1G APOSOR312435-250MG
tert-Butyl (3-aminobicyclo[1.1.1]pent-1-yl)carbamate
tert-Butyl (3-aminobicyclo[1.1.1]pent-1-yl)carbamate
Formula: C₁₀H₁₈N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD27987307
CAS Number: 1638767-25-5