tert-Butyl 3-(bromomethyl)bicyclo[1.1.1]pentane-1-carboxylate
Supplier: Apollo Scientific
Warning
OR312464-1G OR312464-250MG
APOSOR312464-1GEA 3360 EUR
APOSOR312464-1G APOSOR312464-250MG
tert-Butyl 3-(bromomethyl)bicyclo[1.1.1]pentane-1-carboxylate
tert-Butyl 3-(bromomethyl)bicyclo[1.1.1]pentane-1-carboxylate
Formula: C₁₁H₁₇BrO₂
MW: 261.155 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29037328
CAS Number: 138732-34-0