1-Oxa-4,9-diazaspiro[5.5]undecane, N9-BOC protected 97%
Supplier: Apollo Scientific
Warning
Synonyms: tert-Butyl 1-oxa-4,9-diazaspiro[5.5]undecane-9-carboxylate
OR313020-1G OR313020-250MG
APOSOR313020-1GEA 159 EUR
APOSOR313020-1G APOSOR313020-250MG
1-Oxa-4,9-diazaspiro[5.5]undecane, N9-BOC protected 97%
1-Oxa-4,9-diazaspiro[5.5]undecane, N9-BOC protected
Formula: C₁₃H₂₄N₂O₃
MW: 256.341 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11227065
CAS Number: 930785-40-3