1-(3,5-Dichlorophenyl)ethylamine hydrochloride
Supplier: Apollo Scientific
Warning
Synonyms: 1-(3,5-Dichlorophenyl)ethylamine Hydrochloride
OR314005-1G OR314005-250MG OR314005-5G
APOSOR314005-1GEA 150 EUR
APOSOR314005-1G APOSOR314005-250MG APOSOR314005-5G
1-(3,5-Dichlorophenyl)ethylamine hydrochloride
1-(3,5-Dichlorophenyl)ethylamine hydrochloride
Formula: C₈H₉Cl₂N·ClH
MW: 226.531 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18705305
CAS Number: 321318-36-9