tert-Butyl 4-bromobenzylcarbamate 98%

Supplier: Apollo Scientific

OR315540-10G OR315540-25G OR315540-5G
APOSOR315540-10GEA 173 EUR
APOSOR315540-10G APOSOR315540-25G APOSOR315540-5G
tert-Butyl 4-bromobenzylcarbamate 98%
tert-Butyl 4-bromobenzylcarbamate
Formula: C₁₂H₁₆BrNO₂
MW: 286.165 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08703140
CAS Number: 68819-84-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR