2-(Cyclopropyl)-5-methoxybenzenesulphonyl chloride 95%
Supplier: Apollo Scientific
OR315851-1G OR315851-250MG OR315851-5G
APOSOR315851-1GEA 487 EUR
APOSOR315851-1G APOSOR315851-250MG APOSOR315851-5G
2-(Cyclopropyl)-5-methoxybenzenesulphonyl chloride 95%
2-(Cyclopropyl)-5-methoxybenzenesulphonyl chloride
Formula: C₁₀H₁₁ClO₃S
MW: 246.711 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32669831
CAS Number: 2514953-07-0
UN: 3261
ADR: 8 III