5-Cyclopropylpyridine-3-carbaldehyde
Supplier: Apollo Scientific
OR315868-1G OR315868-250MG OR315868-5G
APOSOR315868-1GEA 464 EUR
APOSOR315868-1G APOSOR315868-250MG APOSOR315868-5G
5-Cyclopropylpyridine-3-carbaldehyde
5-Cyclopropylpyridine-3-carbaldehyde
Formula: C₉H₉NO
MW: 147.174 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18255592
CAS Number: 1211589-30-8