tert-Butyl-(R)-(+)-2-hydroxybutyrate
Supplier: Apollo Scientific
Danger
Synonyms: (R)-tert-Butyl 2-hydroxybutanoate
OR316008-250MG OR316008-1G OR316008-5G
APOSOR316008-250MGEA 87 EUR
APOSOR316008-250MG APOSOR316008-1G APOSOR316008-5G
tert-Butyl-(R)-(+)-2-hydroxybutyrate
tert-Butyl-(R)-(+)-2-hydroxybutyrate
Formula: C₈H₁₆O₃
MW: 160.211 g/mol
Melting Pt: 52…54 °C
Storage Temperature: Ambient
MDL Number: MFCD00210055
CAS Number: 206996-51-2