6-Bromo-3-iodo-1H-pyrrolo[3,2-c]pyridine 99%
Supplier: Apollo Scientific
Warning
OR31655-1G OR31655-250MG
APOSOR31655-1GEA 325 EUR
APOSOR31655-1G APOSOR31655-250MG
6-Bromo-3-iodo-1H-pyrrolo[3,2-c]pyridine 99%
6-Bromo-3-iodo-1H-pyrrolo[3,2-c]pyridine
Formula: C₇H₄BrIN₂
MW: 322.929 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09749987
CAS Number: 1000341-73-0