Ethyl 4-(3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenoxy)butanoate 99%
Supplier: Apollo Scientific
Warning
OR321291-250MG
APOSOR321291-250MGEA 139 EUR
APOSOR321291-250MG
Ethyl 4-(3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenoxy)butanoate 99%
Ethyl 4-(3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenoxy)butanoate
Formula: C₁₈H₂₇BO₅
MW: 334.215 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08706023
CAS Number: 850411-09-5