tert-Butyl (S)-3-formylpiperidine-1-carboxylate 97%
Supplier: Apollo Scientific
Warning
OR321300-1G OR321300-250MG
APOSOR321300-1GEA 429 EUR
APOSOR321300-1G APOSOR321300-250MG
tert-Butyl (S)-3-formylpiperidine-1-carboxylate 97%
tert-Butyl (S)-3-formylpiperidine-1-carboxylate
Formula: C₁₁H₁₉NO₃
MW: 213.273 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18633365
CAS Number: 1008562-87-5