tert-Butyl (2-(pyrrolidin-3-yl)ethyl)carbamate 97%
Supplier: Apollo Scientific
Warning
OR321307-1G OR321307-100MG
APOSOR321307-1GEA 1050 EUR
APOSOR321307-1G APOSOR321307-100MG
tert-Butyl (2-(pyrrolidin-3-yl)ethyl)carbamate 97%
tert-Butyl (2-(pyrrolidin-3-yl)ethyl)carbamate
Formula: C₁₁H₂₂N₂O₂
MW: 214.305 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12828830
CAS Number: 169750-93-0