3-Iodo-1H-pyrrolo[3,2-b]pyridine 97%
Supplier: Apollo Scientific
Warning
OR321324-1G OR321324-250MG
APOSOR321324-1GEA 50.9 EUR
APOSOR321324-1G APOSOR321324-250MG
3-Iodo-1H-pyrrolo[3,2-b]pyridine 97%
3-Iodo-1H-pyrrolo[3,2-b]pyridine
Formula: C₇H₅IN₂
MW: 244.032 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11559019
CAS Number: 1083181-26-3