tert-Butyl (R)-4-(1-aminoethyl)piperidine-1-carboxylate 97%
Supplier: Apollo Scientific
Warning
OR321338-1G OR321338-250MG
APOSOR321338-1GEA 330 EUR
APOSOR321338-1G APOSOR321338-250MG
tert-Butyl (R)-4-(1-aminoethyl)piperidine-1-carboxylate 97%
tert-Butyl (R)-4-(1-aminoethyl)piperidine-1-carboxylate
Formula: C₁₂H₂₄N₂O₂
MW: 228.331 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22666109
CAS Number: 1036027-86-7