4,5-Dibromo-3-methoxythiophene-2-carboxylic acid 95%
Supplier: Apollo Scientific
Warning
OR321346-250MG OR321346-1G
APOSOR321346-250MGEA 502 EUR
APOSOR321346-250MG APOSOR321346-1G
4,5-Dibromo-3-methoxythiophene-2-carboxylic acid 95%
4,5-Dibromo-3-methoxythiophene-2-carboxylic acid
Formula: C₆H₄Br₂O₃S
MW: 315.967 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26129618
CAS Number: 120715-53-9