6-Bromo-1H-pyrrolo[3,2-b]pyridine-3-carbaldehyde 97%
Supplier: Apollo Scientific
Warning
OR321373-1G OR321373-5G
APOSOR321373-1GEA 75.4 EUR
APOSOR321373-1G APOSOR321373-5G
6-Bromo-1H-pyrrolo[3,2-b]pyridine-3-carbaldehyde 97%
6-Bromo-1H-pyrrolo[3,2-b]pyridine-3-carbaldehyde
Formula: C₈H₅BrN₂O
MW: 225.042 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12963037
CAS Number: 1190312-27-6