4-(3,4-Dichlorophenyl)piperidine hydrochloride 97%
Supplier: Apollo Scientific
OR321393-1G OR321393-100MG OR321393-250MG
APOSOR321393-1GEA 1010 EUR
APOSOR321393-1G APOSOR321393-100MG APOSOR321393-250MG
4-(3,4-Dichlorophenyl)piperidine hydrochloride 97%
4-(3,4-Dichlorophenyl)piperidine hydrochloride
Formula: C₁₁H₁₃Cl₂N·ClH
MW: 266.595 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08544123
CAS Number: 941711-46-2