(R)-3-(4-Bromophenyl)piperidine (2R,3R)-2,3-dihydroxysuccinate
Supplier: Apollo Scientific
OR321441-1G OR321441-250MG
APOSOR321441-1GEA 493 EUR
APOSOR321441-1G APOSOR321441-250MG
(R)-3-(4-Bromophenyl)piperidine (2R,3R)-2,3-dihydroxysuccinate
(R)-3-(4-Bromophenyl)piperidine (2R,3R)-2,3-dihydroxysuccinate
Formula: C₁₁H₁₄BrN·C₄H₆O₆
MW: 390.226 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD31629926
CAS Number: 2244064-18-2