(R)-3-(4-Bromophenyl)piperidine
Supplier: Apollo Scientific
OR321494-1G OR321494-250MG
APOSOR321494-1GEA 493 EUR
APOSOR321494-1G APOSOR321494-250MG
(R)-3-(4-Bromophenyl)piperidine
(R)-3-(4-Bromophenyl)piperidine
Formula: C₁₁H₁₄BrN
MW: 240.14 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20459850
CAS Number: 1336754-69-8