1-(tert-Butyl) 2-methyl (2S,4R)-4-hydroxypiperidine-1,2-dicarboxylate
Supplier: Apollo Scientific
OR321495-1G OR321495-250MG
APOSOR321495-1GEA 915 EUR
APOSOR321495-1G APOSOR321495-250MG
1-(tert-Butyl) 2-methyl (2S,4R)-4-hydroxypiperidine-1,2-dicarboxylate
1-(tert-Butyl) 2-methyl (2S,4R)-4-hydroxypiperidine-1,2-dicarboxylate
Formula: C₁₂H₂₁NO₅
MW: 259.299 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD02259709
CAS Number: 254882-06-9