(R)-3-(4-Bromophenyl)piperidine hydrochloride
Supplier: Apollo Scientific
OR321496-1G OR321496-250MG
APOSOR321496-1GEA 667 EUR
APOSOR321496-1G APOSOR321496-250MG
(R)-3-(4-Bromophenyl)piperidine hydrochloride
(R)-3-(4-Bromophenyl)piperidine hydrochloride
Formula: C₁₁H₁₄BrN·ClH
MW: 276.601 g/mol
Storage Temperature: Refrigerator
CAS Number: 2409590-07-2