4-(2,3-Dihydrobenzo[b][1,4]dioxin-6-yl)piperidine hydrochloride
Supplier: Apollo Scientific
OR321502-1G OR321502-250MG
APOSOR321502-1GEA 799 EUR
APOSOR321502-1G APOSOR321502-250MG
4-(2,3-Dihydrobenzo[b][1,4]dioxin-6-yl)piperidine hydrochloride
4-(2,3-Dihydrobenzo[b][1,4]dioxin-6-yl)piperidine hydrochloride
Formula: C₁₃H₁₇NO₂·ClH
MW: 255.741 g/mol
Storage Temperature: Refrigerator
CAS Number: 2409597-47-1