2-(3-Methoxyphenyl)piperidine hydrochloride
Supplier: Apollo Scientific
OR321505-1G OR321505-250MG
APOSOR321505-1GEA 681 EUR
APOSOR321505-1G APOSOR321505-250MG
2-(3-Methoxyphenyl)piperidine hydrochloride
2-(3-Methoxyphenyl)piperidine hydrochloride
Formula: C₁₂H₁₇NO·ClH
MW: 227.73 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD02178980
CAS Number: 1187172-83-3