3-(3-Bromophenyl)piperidin-2-one
Supplier: Apollo Scientific
OR321517-1G OR321517-250MG
APOSOR321517-1GEA 957 EUR
APOSOR321517-1G APOSOR321517-250MG
3-(3-Bromophenyl)piperidin-2-one
3-(3-Bromophenyl)piperidin-2-one
Formula: C₁₁H₁₂BrNO
MW: 254.123 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD19358730
CAS Number: 1267325-95-0