3-(2-Bromophenyl)piperidin-2-one
Supplier: Apollo Scientific
OR321518-1G
OR321518-250MG
APOSOR321518-1GEA
1120
EUR
APOSOR321518-1G
APOSOR321518-250MG
3-(2-Bromophenyl)piperidin-2-one
3-(2-Bromophenyl)piperidin-2-one
Formula:
C₁₁H₁₂BrNO MW: 254.123 g/mol Storage Temperature: Refrigerator |
CAS Number:
1784007-12-0 |