5-(Tetrahydro-2H-pyran-4-yl)pyridin-2-amine
Supplier: Apollo Scientific
OR321527-1G OR321527-250MG
APOSOR321527-1GEA 754 EUR
APOSOR321527-1G APOSOR321527-250MG
5-(Tetrahydro-2H-pyran-4-yl)pyridin-2-amine
5-(Tetrahydro-2H-pyran-4-yl)pyridin-2-amine
Formula: C₁₀H₁₄N₂O
MW: 178.231 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12033508
CAS Number: 1159814-73-9