Methyl 4-(piperidin-2-yl)benzoate hydrochloride
Supplier: Apollo Scientific
OR321532-1G OR321532-250MG
APOSOR321532-1GEA 719 EUR
APOSOR321532-1G APOSOR321532-250MG
Methyl 4-(piperidin-2-yl)benzoate hydrochloride
Methyl 4-(piperidin-2-yl)benzoate hydrochloride
Formula: C₁₃H₁₇NO₂·ClH
MW: 255.741 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD02179145
CAS Number: 863769-42-0