(R)-4-(Piperidin-3-yl)aniline
Supplier: Apollo Scientific
OR321544-1G OR321544-250MG
APOSOR321544-1GEA 1210 EUR
APOSOR321544-1G APOSOR321544-250MG
(R)-4-(Piperidin-3-yl)aniline
(R)-4-(Piperidin-3-yl)aniline
Formula: C₁₁H₁₆N₂
MW: 176.258 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20458031
CAS Number: 1334823-70-9