2,5-Dimethyl-6-phenylpyrazolo[1,5-a]pyrimidin-7-amine 95%
Supplier: Apollo Scientific
OR32244-1G OR32244-5G
APOSOR32244-1GEA 180 EUR
APOSOR32244-1G APOSOR32244-5G
2,5-Dimethyl-6-phenylpyrazolo[1,5-a]pyrimidin-7-amine 95%
2,5-Dimethyl-6-phenylpyrazolo[1,5-a]pyrimidin-7-amine
Formula: C₁₄H₁₄N₄
MW: 238.288 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02661827
CAS Number: 428845-73-2