1-Isopropyl-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride
Supplier: Apollo Scientific
Warning
OR322463-250MG OR322463-1G
APOSOR322463-250MGEA 46.4 EUR
APOSOR322463-250MG APOSOR322463-1G
1-Isopropyl-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride
1-Isopropyl-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride
Formula: C₁₄H₂₁NO₂·ClH
MW: 271.783 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03095694
CAS Number: 214046-76-1