2-(3-Methylpyridin-2-yl)ethanamine
Supplier: Apollo Scientific
OR322555-250MG OR322555-500MG
APOSOR322555-250MGEA 328 EUR
APOSOR322555-250MG APOSOR322555-500MG
2-(3-Methylpyridin-2-yl)ethanamine
2-(3-Methylpyridin-2-yl)ethanamine
Formula: C₈H₁₂N₂
MW: 136.194 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07367941
CAS Number: 851670-19-4