4-(2-Methylpyridin-3-yloxy)-6-chloro-5-Methylpyrimidine 97%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Chloro-5-methyl-6-((2-methylpyridin-3-yl)oxy)pyrimidine
OR322841-250MG OR322841-1G
APOSOR322841-250MGEA 92.8 EUR
APOSOR322841-250MG APOSOR322841-1G
4-(2-Methylpyridin-3-yloxy)-6-chloro-5-Methylpyrimidine 97%
4-(2-Methylpyridin-3-yloxy)-6-chloro-5-Methylpyrimidine
Formula: C₁₁H₁₀ClN₃O
MW: 235.67 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12756519
CAS Number: 930093-72-4