Methyl 4'-hydroxy-[1,1'-biphenyl]-4-carboxylate 95%
Supplier: Apollo Scientific
OR32319-1G OR32319-5G
APOSOR32319-1GEA 143 EUR
APOSOR32319-1G APOSOR32319-5G
Methyl 4'-hydroxy-[1,1'-biphenyl]-4-carboxylate 95%
Methyl 4'-hydroxy-[1,1'-biphenyl]-4-carboxylate
Formula: C₁₄H₁₂O₃
MW: 228.243 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01814566
CAS Number: 40501-41-5