2,3-Dimethoxy-6H,7H,8H,9H,10H,12H-azepino[2,1-b]quinazolin-12-one 95%
Supplier: Apollo Scientific
OR32496-1G OR32496-5G
APOSOR32496-1GEA 339 EUR
APOSOR32496-1G APOSOR32496-5G
2,3-Dimethoxy-6H,7H,8H,9H,10H,12H-azepino[2,1-b]quinazolin-12-one 95%
2,3-Dimethoxy-6H,7H,8H,9H,10H,12H-azepino[2,1-b]quinazolin-12-one
Formula: C₁₅H₁₈N₂O₃
MW: 274.315 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03617945
CAS Number: 475638-35-8