(1E)-1-(2-Phenylhydrazin-1-ylidene)propan-2-one 95%
Supplier: Apollo Scientific
OR32758-1G OR32758-5G
APOSOR32758-1GEA 209 EUR
APOSOR32758-1G APOSOR32758-5G
(1E)-1-(2-Phenylhydrazin-1-ylidene)propan-2-one 95%
(1E)-1-(2-Phenylhydrazin-1-ylidene)propan-2-one
Formula: C₉H₁₀N₂O
MW: 162.189 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00026242
CAS Number: 27843-96-5