(2S)-4-Iodo-2-methyl-2,3-dihydro-1H-pyrrolo[2,3-b]pyridine 95%
Supplier: Apollo Scientific
Warning
OR33656-1G OR33656-250MG
APOSOR33656-1GEA 766 EUR
APOSOR33656-1G APOSOR33656-250MG
(2S)-4-Iodo-2-methyl-2,3-dihydro-1H-pyrrolo[2,3-b]pyridine 95%
(2S)-4-Iodo-2-methyl-2,3-dihydro-1H-pyrrolo[2,3-b]pyridine
Formula: C₈H₉IN₂
MW: 260.075 g/mol
Storage Temperature: Ambient
CAS Number: 2366997-19-3