3-Amino-5-bromo-2-nitropyridine 95%
Supplier: Apollo Scientific
Warning
Synonyms: 5-Bromo-2-nitropyridin-3-amine
OR345345-1G OR345345-250MG OR345345-5G
APOSOR345345-1GEA 98.6 EUR
APOSOR345345-1G APOSOR345345-250MG APOSOR345345-5G
3-Amino-5-bromo-2-nitropyridine 95%
3-Amino-5-bromo-2-nitropyridine
Formula: C₅H₄BrN₃O₂
Storage Temperature: Refrigerator
MDL Number: MFCD11040269
CAS Number: 433226-05-2