Methyl 5-hydroxymethyl-1-methyl-1H-pyrazole-3-carboxylate
Supplier: Apollo Scientific
Warning
OR346081-1G OR346081-5G
APOSOR346081-1GEA 243 EUR
APOSOR346081-1G APOSOR346081-5G
Methyl 5-hydroxymethyl-1-methyl-1H-pyrazole-3-carboxylate
Methyl 5-hydroxymethyl-1-methyl-1H-pyrazole-3-carboxylate
Formula: C₇H₁₀N₂O₃
MW: 170.166 g/mol
Melting Pt: 84…87 °C
Storage Temperature: Ambient
MDL Number: MFCD10568328
CAS Number: 1208081-63-3