5-(3-Chloro-phenyl)-3-methyl-isoxazole-4-carboxylic acid
Supplier: Apollo Scientific
Warning
OR346227-1G OR346227-5G
APOSOR346227-1GEA 0 EUR
APOSOR346227-1G APOSOR346227-5G
5-(3-Chloro-phenyl)-3-methyl-isoxazole-4-carboxylic acid
5-(3-Chloro-phenyl)-3-methyl-isoxazole-4-carboxylic acid
Formula: C₁₁H₈ClNO₃
MW: 237.639 g/mol
Melting Pt: 176…178 °C
Storage Temperature: Ambient
MDL Number: MFCD11584333
CAS Number: 92545-96-5