2-Acetyl-3-methoxy-but-2-enoic acid methyl ester
Supplier: Apollo Scientific
OR346251-1G OR346251-500MG OR346251-5G
APOSOR346251-1GEA 0 EUR
APOSOR346251-1G APOSOR346251-500MG APOSOR346251-5G
2-Acetyl-3-methoxy-but-2-enoic acid methyl ester
2-Acetyl-3-methoxy-but-2-enoic acid methyl ester
Formula: C₈H₁₂O₄
MW: 172.178 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19981463
CAS Number: 943910-00-7