5-Bromomethyl-1-methyl-1H-pyrazole-3-carboxylic acid methyl ester
Supplier: Apollo Scientific
Danger
OR346296-1G OR346296-250MG
APOSOR346296-1GEA 411 EUR
APOSOR346296-1G APOSOR346296-250MG
5-Bromomethyl-1-methyl-1H-pyrazole-3-carboxylic acid methyl ester
5-Bromomethyl-1-methyl-1H-pyrazole-3-carboxylic acid methyl ester
Formula: C₇H₉BrN₂O₂
MW: 233.063 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21362488
CAS Number: 878744-22-0