2-Dimethylamino-4,6-dimethyl-nicotinonitrile
Supplier: Apollo Scientific
OR346299-1G OR346299-5G
APOSOR346299-1GEA 0 EUR
APOSOR346299-1G APOSOR346299-5G
2-Dimethylamino-4,6-dimethyl-nicotinonitrile
2-Dimethylamino-4,6-dimethyl-nicotinonitrile
Formula: C₁₀H₁₃N₃
MW: 175.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01005832
CAS Number: 1233123-61-9