2,4-Dichloro-5-formylnicotinonitrile 95+%
Supplier: Apollo Scientific
OR346410-1G OR346410-500MG
APOSOR346410-1GEA 318 EUR
APOSOR346410-1G APOSOR346410-500MG
2,4-Dichloro-5-formylnicotinonitrile 95+%
2,4-Dichloro-5-formylnicotinonitrile
Formula: C₇H₂Cl₂N₂O
MW: 201.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12131124
CAS Number: 176433-56-0