3-Bromo-azepan-2-one 95+%

Supplier: Apollo Scientific

OR346532-1G OR346532-5G
APOSOR346532-1GEA 192 EUR
APOSOR346532-1G APOSOR346532-5G
3-Bromo-azepan-2-one 95+%
3-Bromo-azepan-2-one
Formula: C₆H₁₀BrNO
Storage Temperature: Ambient
MDL Number: MFCD02683824
CAS Number: 3457-66-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR