1-tert-Butyl 7-methyl 1H-indole-1,7-dicarboxylate 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1-(tert-Butoxycarbonyl)-7-(methoxycarbonyl)-1H-indole
OR3492-1G OR3492-5G
APOSOR3492-1GEA 29 EUR
APOSOR3492-1G APOSOR3492-5G
1-tert-Butyl 7-methyl 1H-indole-1,7-dicarboxylate 98%
1-tert-Butyl 7-methyl 1H-indole-1,7-dicarboxylate
Formula: C₁₅H₁₇NO₄
MW: 275.3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09027276
CAS Number: 917562-23-3