1-Benzyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2,3,6-tetrahydropyridine
Supplier: Apollo Scientific
OR350073-1G OR350073-5G
APOSOR350073-1GEA 104 EUR
APOSOR350073-1G APOSOR350073-5G
1-Benzyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2,3,6-tetrahydropyridine
1-Benzyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2,3,6-tetrahydropyridine
Formula: C₁₈H₂₆BNO₂
Storage Temperature: Ambient
MDL Number: MFCD11501654
CAS Number: 1048976-83-5